Learn More
Medchemexpress LLC HY-N4327 1mg Medchemexpress, Eurycomalactone CAS:23062-24-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN43271MG
Encompass_Preferred
Medchemexpress, HY-N4327 1mg Eurycomalactone CAS:23062-24-0 Eurycomalactone is a natural product found in Eurycoma longifolia Jack., acts as a potent NF-κB inhibitor, with an IC50 of 0.5 μM. Eurycomalactone inhibits protein synthesis, depletes cyclin D1, but does not affect TNFα-induced degradation of IκBα or the phosphorylation of IKKα/β and IκBα. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2935