Learn More
Apexbio Technology LLC Eurycomanone 84633-29-4 5mg
SDP
Supplier:  Apexbio Technology LLC BA81605
Encompass_Preferred
Eurycomanone (CAS 84633-29-4) is a quassinoid compound studied for its modulatory effects on steroidogenesis It exerts its biological activity by inhibiting phosphodiesterase and aromatase enzymes thereby interfering with estrogen biosynthesis pathways Through this dual enzyme inhibition eurycomanone has been shown to enhance spermatogenesis in relevant research models This small molecule is utilized in biomedical studies investigating male reproductive function and the regulation of hormone synthesis
Catalog No. 50-000-89522