Learn More
Medchemexpress LLC Eurycomanone | 84633-29-4 | MFCD31726288 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN50121MG
Encompass_Preferred
Eurycomanone, also known as Pasakbumin A, is a natural quassinoid that increases spermatogenesis by inhibiting the activity of phosphodiesterase and aromatase in steroidogenesis. It has been shown to significantly increase testosterone production dose-dependently. Molecular docking studies indicate that Eurycomanone binds at different sites than some other inhibitors and can improve inhibition of aromatase in reducing estrogen production. This product is intended for research and analytical applications.
  • Increases spermatogenesis
  • Inhibits phosphodiesterase and aromatase
  • Significantly increases testosterone production
  • Suitable for research and analytical applications
  • Demonstrates improved inhibition of aromatase in reducing estrogen production
Catalog No. 50-004-18659