Learn More
Cayman Chemical EuscaphIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3616210 MG
Encompass_Preferred
A triterpene with diverse biological activities; inhibits the proliferation of CNE-1 and C666-1 nasopharyngeal carcinoma cells at 5 and 10 UG/ml; inhibits AChE (IC50 = 35.9 µM) and α-glucosidase (IC50 = 24.9 µM); reduces the production of NO and levels of iNOS and COX-2 in LPS-stimulated RAW 264.7 cells; reduces serum IgE and IgG2a levels, EAr tissue mast cell infiltration, and pruritis in a mouse model of difluoroethane- and 2,4-dinitrochlorobenzene-induced atopic dermatitis at 10 mg/kg
Catalog No. 50-290-9188