Learn More
Biotium EV stains Cell and Organelle Stains Exosome EV stains ExoBrite 645 675 1/EA
Supplier:  Biotium 30137
Encompass_Preferred
ExoBrite 645/675 True EV Membrane Stain is a membrane binding dye that stains EVs with full coverage and broad compatibility These dyes have improved solubility resulting in lower background and better signal to noise in EV detection compared to other membrane dyes 500X in DMSO 500 uL
Catalog No. 50-005-01521