Learn More
Biotium EV stains Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates ExoBrite 560 585 1/EA
Supplier:  Biotium 30125T
Encompass_Preferred
ExoBrite WGA EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Wheat Germ Agglutinin (WGA) binds to carbohydrates in the EV membrane for bright staining Ex/Em 562/584 nm 100 labelings
Catalog No. 50-005-01104