Learn More
Glas Col LLC EVAPORATOR SMALL SYS 24 POS
SDP
Supplier:  Glas Col LLC 109A YH1
Encompass_Preferred
  • Efficient solvent removal utilizing a nitrogen blow-down method for rapid and controlled evaporation.
  • Uniform evaporation ensuring complete equal flow rates from all gas ports to prevent overheating of sensitive compounds.
  • Versatile sample accommodation with a single manifold design compatible with various standard vial sizes.
  • Precise control with an integrated flowmeter and heating capability up to 100 °C.
  • Enhanced workflow with easy closure of individual gas ports to save nitrogen and a high-quality jacking stand for needle adjustments.
Catalog No. NC3626707