Learn More
Medchemexpress LLC Evernic acid | 537-09-7 | 99.7% | 10 MG

Supplier: Medchemexpress LLC HY12136210MG
Evernic acid is a lichen-derived secondary metabolite reported to exhibit anticancer, antifungal, antimicrobial, neuroprotective, and anti-inflammatory activities. It is supplied as a research-grade small molecule for laboratory use and intended for research use only.
- Research-grade lichen-derived secondary metabolite.
- High stated purity of 99.7%.
- Molecular formula C17H16O7 and molecular weight 332.30 g/mol.
- Available in 5 mg and 10 mg pack sizes; larger sizes available by quote.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.