Learn More
Medchemexpress LLC Everolimus | 159351-69-6 | MFCD00929329 | 99.7% | 958.22 g/mol | C53H83NO14 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY10218200MG
Encompass_Preferred
Everolimus is a rapamycin-derivative mTORC1 inhibitor used in research to modulate cell growth, proliferation, autophagy, and immune responses. It binds FKBP12 to form an inhibitory complex with mTOR and is commonly used in cellular and preclinical studies.
  • Potent, selective mTORC1 inhibitor.
  • Binds FKBP12 to form an inhibitory complex with mTOR.
  • High purity reported (99.74%).
  • Available in multiple research pack sizes up to 100 mg and as a 10 mM solution.
  • Intended for research and analytical applications.
Catalog No. 50-002-46576