Learn More
Selleck Chemical LLC Everolimus S1120-1g
SDP
Supplier:  Selleck Chemical LLC S11201G
Encompass_Preferred
Everolimus is an mTOR inhibitor of FKBP12 with IC50 of 1 6-2 4 nM in a cell-free assay Everolimus induces cell apoptosis and autophagy and inhibits tumor cells proliferation
Catalog No. 50-310-8531