Learn More
Medchemexpress LLC Everolimus-d4 | 1338452-54-2 | MFCD21363580 | 98.7% | 962.25 | C53H79D4NO14 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY10218S1MG
Encompass_Preferred
Everolimus-d4 is the deuterium-labeled analogue of everolimus used as a stable isotope internal standard for quantitative mass spectrometry and for research into mTOR-related biology.
  • Deuterium-labeled internal standard for LC-MS and GC-MS quantification of everolimus.
  • High analytical purity suitable for quantitative assays.
  • Available in small research-scale quantities (e.g., 500 μg, 1 mg, 5 mg, 10 mg).
  • Molecular formula C53H79D4NO14 and molecular weight ≈962.25 g/mol.
  • Intended for research use only; not for human, clinical, or diagnostic use.
Catalog No. 50-002-46575