Learn More
Medchemexpress LLC Evifacotrep | 2413739-88-3 | 99.7% | C18H12ClF4N5O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13281310MG
Encompass_Preferred
Evifacotrep is a small-molecule antagonist of transient receptor potential channels TRPC5 and TRPC4, provided as a white to off-white solid (CAS 2413739-88-3) for in vitro and preclinical research into neurological and ion-channel biology.
  • Potent TRPC5/TRPC4 antagonist with IC50 values ≤50 nM.
  • High purity (99.7%) as reported by the supplier certificate of analysis.
  • Soluble in DMSO (50 mg/mL) with ultrasonic warming recommended for dissolution.
  • Stable as powder at -20°C for up to 3 years under recommended storage.
  • Suitable for biochemical and cellular assay applications targeting TRP channels.
Catalog No. 50-002-58722