Learn More
TARGETMOL CHEMICALS INC EVOBRUTINIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T438710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Evobrutinib(M2951) also known as M-2951 and MSC-2364447C is a highly selective inhibitor of the Bruton's tyrosine kinase (BTK) which is important in the development and functioning of various immune cells including B -lymphocytes and macrophages. PreClinical research suggests it may be therapeutically useful in certain autoimmune diseases. purity: 99%
Catalog No. 50-257-9890