Learn More
Medchemexpress LLC Evocalcet | 870964-67-3 | 99.3% | 374.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY17613
Encompass_Preferred
Evocalcet is an orally active calcium sensing receptor (CaSR) agonist. It inhibits the secretion of parathyroid hormone (PTH) from parathyroid gland cells and is suitable for research related to hyperparathyroidism.
  • Orally active calcium sensing receptor (CaSR) agonist
  • Inhibits parathyroid hormone (PTH) secretion from parathyroid gland cells
  • Increases intracellular calcium ([Ca2+]i) in hCaR-HEK293 cells with an EC50 of 92.7 nM
  • Does not significantly inhibit cytochrome P450 (CYP) isozymes
  • Affects serum parathyroid hormone and calcium levels in vivo
  • Reduces serum parathyroid hormone and calcium levels and increases inorganic phosphorus levels in animal models of secondary hyperparathyroidism
  • Used for research of hyperparathyroidism
Catalog No. 50-003-93318