Learn More
Cayman Chemical -EvodIamIn 250mg
SDP
Supplier:  Cayman Chemical 16885250 MG
Encompass_Preferred
A natural indole alkaloid with demonstrated beneficial effects in cancer, obesity, inflammation, and many other conditions; an AhR antagonist (Ki = 28 nM), activator of TRPV1 (EC50 = 45 nM), and inhibitor of signaling through NF-κB
Catalog No. 50-296-6056