Learn More
Medchemexpress LLC HY-10535 5mg Medchemexpress, Evofosfamide CAS:918633-87-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105355MG
Encompass_Preferred
Medchemexpress, HY-10535 5mg Evofosfamide CAS:918633-87-1 Evofosfamide (TH-302) is a hypoxia-activated prodrug with IC50 of 10 μM and 1000 μM in hypoxia (N2) and normoxia (21% O2), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8271