Learn More
Medchemexpress LLC HY-117985B 5mg Medchemexpress, Evogliptin (tartrate) CAS:1222102-51-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY117985B5MG
Encompass_Preferred
Medchemexpress, HY-117985B 5mg Evogliptin (tartrate) CAS:1222102-51-3 Evogliptin tartrate is a potent, orally bioavailable and selective dipeptidyl peptidase-4 (DPP-4) inhibitor, with antidiabetic activity. Evogliptin tartrate has potential for anti-atherosclerosis therapy that targets arterial inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0912