Learn More
Electron Microscopy Sciences Ex Wide Compact Locking 4 Drawer Cart with Red Drawers
SDP
Supplier:  Electron Microscopy Sciences 6438659
Encompass_Preferred
White Polyethylene Extra Wide Compact Locking 4 Drawer Cart is made of strong, durable, and chemically resistant white polyethylene plastic construction. It features has 4 locking drawers 3 inches deep and at 27 inches fits under most counters, tables, desks and benches. Push-pull handle is built in for easy mobility as it rolls on heavy duty casters with brakes. All drawers lock at once with two keys included.
Color: Red Drawers
Catalog No. 50-192-7297