Learn More
Hanna Pharmaceutical Supply Co., Inc. EXACTAMED Oral Dispenser 60 mL
Supplier:  Hanna Pharmaceutical Supply Co., Inc. H9388160
Encompass_Preferred
EXACTAMED Oral Dispenser, 60 mL, Amber, Non-Luer Slip, Black O-Ring, Self Righting Tip Cap Included. Pharmacy Pack 25/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
Catalog No. NC1847251