Learn More
Medchemexpress LLC Exarafenib | 2639957-39-2 | 99.7% | 521.58 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14726810MM/1ML
Encompass_Preferred
Exarafenib is an orally-available, selective pan-RAF inhibitor that is effective in RAF-dependent cancers, including all classes of BRAF alterations. It suppresses MAPK signaling in RAF-dependent melanoma cell lines and demonstrates anticancer activity.
  • Orally-available, selective pan-RAF inhibitor
  • Effective against RAF-dependent cancers, including BRAF alterations
  • Suppresses MAPK signaling in RAF-dependent melanoma cell lines
  • Demonstrates anticancer activity
Catalog No. 50-003-62487