Learn More
Medchemexpress LLC Exatecan-d5 mesylate | 2819276-88-3 | 99.8% | 536.58 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13631AS50MG
Encompass_Preferred
Exatecan-d5 mesylate is a deuterium-labeled version of Exatecan mesylate. It acts as a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL), and is used in cancer research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis using NMR, GC-MS, or LC-MS.
Catalog No. 50-003-48301