Learn More
Medchemexpress LLC Exatecan Intermediate 1-d5 | 1346617-23-9 | 99.90% | 268.28 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY42487S
Encompass_Preferred
Exatecan Intermediate 1-d5 is the deuterium labeled Exatecan Intermediate 1. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-02366