Learn More
Medchemexpress LLC Exatecan intermediate 12 | 110351-93-4 | C15H17NO6 | 5 G
SDP
Supplier:  Medchemexpress LLC HY451715G
Encompass_Preferred
Exatecan intermediate 12 is a chemical substance used in laboratory research and for the manufacture of other substances. This solid compound has a molecular weight of 307.30 and is classified with acute oral toxicity, skin and serious eye irritation, and may cause respiratory irritation. Proper handling and storage are crucial for safety and stability.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Stable under recommended storage conditions
  • Recommended storage: -20°C for 3 years (powder), 4°C for 2 years (powder)
  • Avoid inhalation, contact with eyes and skin
  • Use in areas with appropriate exhaust ventilation
Catalog No. 50-003-58210