Learn More
Medchemexpress LLC Exatecan Intermediate 6 | 143655-58-7 | 98.8% | 235.26 | 5 G
SDP
Supplier:  Medchemexpress LLC HY443695G
Encompass_Preferred
Exatecan Intermediate 6 is a key intermediate in the synthesis of Exatecan, a potent DNA topoisomerase I inhibitor with an IC50 value of 2.2 μM (0.975 μg/mL) used in cancer research. This compound can also be employed to synthesize Antibody-Drug Conjugates (ADCs). Its appearance is a solid, off-white to light yellow in color.
  • Utilized in cancer research.
  • Can be used to synthesize antibody-drug conjugates.
  • Appears as a solid, off-white to light yellow in color.
Catalog No. 50-004-05116