Learn More
Medchemexpress LLC Exatecan mesylate (1R,9R) | 2938875-39-7 | 99.9% | 531.55 g/mol | C25H26FN3O7S | 5 MG

Supplier: Medchemexpress LLC HY13631J5MG
Exatecan mesylate (1R,9R) is the (1R,9R) stereoisomer of exatecan mesylate, a DNA topoisomerase I inhibitor commonly used as an ADC cytotoxin building block in biomedical research. The compound is supplied as a solid with high enantiomeric and chemical purity and is formulated for use in medicinal chemistry, biological assays, and antibody-drug conjugate development.
- High chemical purity (99.94%).
- Available in small research quantities (5 mg) and larger sizes.
- Soluble in DMSO (16.67 mg/mL) with ultrasonic assistance.
- Solid form; store at 4°C under nitrogen away from moisture.
- Stable in solution when stored at -80°C (up to 6 months) or -20°C (up to 1 month).
- Functional as a DNA topoisomerase I inhibitor and ADC payload building block.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.