Learn More
Cayman Chemical Exatecnmesylate 10mg
SDP
Supplier:  Cayman Chemical 3545210 MG
Encompass_Preferred
A DNA topoisomerase I inhibitor (IC50 = 0.975 UG/ml); inhibits cell growth in a panel of 32 breast, colon, stomach, and lung cancer cell lines (GI50s = 0.125-10.4 ng/ml); reduces tumor growth in an SC-6-JCK gastric adenocarcinoma mouse xenograft model from 3.325-50 mg/kg; reduces tumor weight in HCT116, PC-6, and PC12 mouse xenograft models at 75 mg/kg; has been used in the synthesis of cytotoxic payload moieties of ADCs
Catalog No. 50-290-9603