Learn More
Med Vet International Exel Syringe, 50-60mL, Eccentric Luer Slip, 25/Box, 26301
Supplier:  Med Vet International 23601
Encompass_Preferred
Exel International 50-60mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 50-60mL syringe fits conventional and safety needles. This quantity is 25/BX
Catalog No. NC1741049