Learn More
Cole-Parmer Traceable Excursion-Trac™ Datalogging Thermometer with Calibration; 1 Plastic Bottle Probe
Supplier:  Cole-Parmer 9876767
Encompass_Preferred
Rolling memory retains most recent 525,600 temperature observationsIncludes NIST-traceable certificate from our A2LA-accredited (1750.01) calibration laboratoryMemory can be cleared after downloading data and alarm eventsSet limits for and store up to 10 unique alarm eventsSmart-Alarm™ with visual/audible alarm continues to alert even if conditions return to normal rangeTriple-display—shows high, low, and current
Catalog No. NC3054799