Learn More
Medchemexpress LLC Exendin-3 | 130357-25-4 | 98.3% | 4202.57 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP15435MG
Encompass_Preferred
Exendin-3 is a biologically active peptide isolated from the venoms of the Gila monster lizards, *Heloderma horridum*. It interacts with vasoactive intestinal peptide (VIP) receptors and a newly described putative exendin receptor on dispersed acini from guinea pig pancreas. At high concentrations (>100 nM), it causes a significant increase in cAMP and stimulates amylase release. At lower concentrations (0.1-3 nM), it interacts with the putative exendin receptor, leading to a smaller increase in cAMP.
  • Interacts with VIP receptors and a putative exendin receptor
  • Isolated from *Heloderma horridum* venoms
  • Causes an increase in cAMP and stimulates amylase release
Catalog No. 50-004-24340