Learn More
Medchemexpress LLC Exendin-3/4 (64-86) | 98.29% | 2409.75 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP14475MG
Encompass_Preferred
Exendin-3/4 (64-86) is a polypeptide derived from patent CN 106029087 A. The incretin receptor ligands are obtained from multiple skin toxicity Shan exosomes. This product appears as a white to off-white solid and is intended for research use only.
  • Polypeptide sequence: EAVRLFIEWLKNGGPSSGAPPPS
  • White to off-white solid appearance
  • Intended for research use only
Catalog No. 50-004-23280