Learn More
CPC Scientific H-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2 0.5MG
SDP
Catalog No. 502109267
Encompass
Potent glucagon-like peptide (GLP-1) receptor antagonist.SEQUENCE: H-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2ONE-LETTER SEQUENCE: EGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2MOLECULAR FORMULA: C176H272N46O58S1MOLECULAR WEIGHT: 3992.46STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [196109-31-6]SYNONYMS: RESEARCH AREA: DiabetesREFERENCES:SKU(s): EXED-004A, EXED-004B
Catalog No. 50-210-9267 Supplier CPC Scientific Supplier No. EXED004A
Add to Cart
Add to Cart
Potent glucagon-like peptide (GLP-1) receptor antagonist.SEQUENCE: H-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2ONE-LETTER SEQUENCE: EGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2MOLECULAR FORMULA: C176H272N46O58S1MOLECULAR WEIGHT: 3992.46STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [196109-31-6]SYNONYMS: RESEARCH AREA: DiabetesREFERENCES:SKU(s): EXED-004A, EXED-004B