Learn More
Carbosynth LLC. Exendin-4, 1mg, Sequence originally isolated from venom derived from the modified salivary glands of the Gila monster (Heloderma suspectum)., MW: (M.W. 4186.66), Molecular formula: C184H282N50O60S, CAS# [141758-74-9]
SDP
Supplier:  Carbosynth LLC. PEX3784PI1MG
Encompass_Preferred
Synonyms: H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2, exenatide, H-HGEGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2, Application: GLP-1 (Glucagon-Like Peptide-1) Receptor Agonist, Storage Temperature: -20°C, References: R Göke, et al., J Biol Chem., 26, 19650 (1993)., B. Thorens, et al., Diabetes, 42, 1678, (1993)., A. Alcántara, et al., Arch Biochem Biophys., 341, 1, (1997).
Catalog No. 50-168-6481