Learn More
Bachem Exenatide Acetate
SDP
Supplier:  Bachem 4019602.0500
Encompass_Preferred
Exenatide Acetate, H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2, 141758-74-9, C184H282N50O60S, Mr 4186.63, 0.5mg. Exendin-4. Exendin-4 (exenatide), originally isolated from Heloderma suspectum venom, differs from exendin-3 by only 2 N-terminal amino acid substitutions. This polypeptide causes an increase in acinar cAMP without stimulating amylase release. Exendin-4 and glucagon-like peptide 1 (7-36) amide bind with similar affinity to the glucagon-like peptide 1 receptor. Moreover, exendin-4 is also considered for clinical use in the treatment of type 2 diabetes.
Catalog No. 50-194-6883