Learn More
Cayman Chemical ExendIn-43-39 amIdetrIfl 1mg
SDP
Supplier:  Cayman Chemical 369121 MG
Encompass_Preferred
A GLP-1R agonist and active metabolite of exendin-4 (48-86) amide; induces cAMP accumulation in HEK293 cells expressing human GLP-1R (EC50 = 9 nM)
Catalog No. 50-291-4950