Learn More
Medchemexpress LLC Exendin (5-39) | 196109-27-0 | 99.4% | 3806.30 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP24971MG
Encompass_Preferred
Exendin (5-39) is a potent antagonist of the glucagon-like peptide 1 (GLP-1) receptor. It has been shown to improve memory impairment in rats treated with β-amyloid protein.
  • Potent glucagon-like peptide 1 (GLP-1) receptor antagonist.
  • Improves memory impairment in β-amyloid protein-treated rats.
  • Increases GLT-1 protein levels in the hippocampus of male Wistar rats (in vivo study).
  • Decreases fEPSP decay time and increases the input-output relation in the dentate gyrus (in vivo study).
  • Inhibits long-term depression in the dentate gyrus (in vivo study).
  • Available in various quantities: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
  • Purity of 99.43%.
Catalog No. 50-004-24243