Learn More
Medchemexpress LLC Exendin (9-39) amide | 133514-43-9 | 99.8% | 3369.76 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP0264500UG
Encompass_Preferred
Exendin(9-39) amide, also known as Avexitide, is a glucagon-like peptide-1 (GLP-1) receptor antagonist. It works by competing with endogenous GLP-1 for binding to GLP-1 receptors, effectively counteracting excessive GLP-1 secretion. This product is ideal for research into conditions such as postoperative hypoglycemia (PBH).
  • Plays a role in the control of fasting glucose as a specific GLP-1 receptor antagonist.
  • Raises fasting blood glucose levels in SUR-1 -/- mice when continuously infused subcutaneously.
Catalog No. 50-004-26013