Learn More
Medchemexpress LLC Exendin(9-39) amide (Avexitide) | 133514-43-9 | 99.8% | 3369.76 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP02645MG
Encompass_Preferred
Exendin(9-39) amide (Avexitide) is a glucagon-like peptide-1 (GLP-1) antagonist that competes with endogenous GLP-1 for binding to GLP-1 receptors, thereby antagonizing the effects of excess GLP-1 secretion. It can be used to study postoperative hypoglycemia (PBH) and plays a role in the control of fasting glucose.
  • Acts as a specific GLP-1 receptor antagonist
  • Antagonizes effects of excess GLP-1 secretion
  • Useful for studying postoperative hypoglycemia
  • Elevates fasting blood glucose levels in SUR-1 -/- mice
Catalog No. 50-004-25953