Learn More
Medchemexpress LLC exo-BCN-NHS carbonat 100mg
SDP
Supplier:  Medchemexpress LLC HYW096149100MG
Encompass_Preferred
exo-BCN-NHS carbonate carbonate contains BCN group Strain-promoted alkyne-azide cycloaddition (SPAAC) can also occur with molecules containing DBCO or BCN groups
Catalog No. 50-002-06014