Learn More
New England Biolabs, Inc. Exo-CIP Rapid PCR Cleanup Kit – 400 reactions
SDP
Supplier:  New England Biolabs, Inc. E1050L
Encompass
The Exo-CIP Rapid PCR Cleanup Kit contains optimized formulations of thermolabile Exonuclease I and thermolabile Calf Intestinal Phosphatase (CIP) and is used to rapidly degrade residual PCR primers and dephosphorylate excess dNTPs after amplification. Degradation occurs in only 4 minutes at 37C, and is immediately followed by rapid inactivation of the enzymes by heating for 1 minute at 80C. In just 5 minutes, the PCR product is ready for downstream analysis, such as Sanger Sequencing, SNP detection, or library preparation for NGS. The Exo-CIP Rapid PCR Cleanup Kit is compatible with all commonly-used PCR buffers.
Catalog No. 50-195-9350