Learn More
Cayman Chemical exo-THCP methyl ether 100ug
SDP
Supplier:  Cayman Chemical 40539100 UG
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; intended for research and forensic applications
Catalog No. 50-294-2923