Learn More
Medchemexpress LLC Exo2 | 304684-77-3 | 99.57% | 354.43 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13797510MM/1ML
Encompass_Preferred
Exo2 is a secretion inhibitor that perturbs the trafficking of Shiga toxin between endosomes and the trans-Golgi network. It blocks secretory cargo exit from the ER (endoplasmic reticulum) and disrupts the Golgi apparatus, but does not affect the morphology of the TGN (trans-Golgi network). Exo2 can also stimulate calcium-dependent exocytosis in permeabilized adrenal chromaffin cells.
  • Perturbs Shiga toxin trafficking
  • Blocks secretory cargo exit from ER
  • Disrupts the Golgi apparatus
  • Stimulates calcium-dependent exocytosis
Catalog No. 50-003-48161