Learn More
Medchemexpress LLC Exodus-2/CCL21 Huma 100ug

Supplier: Medchemexpress LLC HYP7166100UG
Recombinant human CCL21 (Exodus-2), corresponding to residues S24-P134, supplied lyophilized for use in bioactivity assays such as chemotaxis. The protein is expressed in E. coli, demonstrates >95% purity by reducing SDS-PAGE, and shows chemotactic activity on human lymphocytes with an ED50 of 11.61 ng/mL.
- Provided as lyophilized powder for long-term storage.
- Contains residues S24-P134 of human CCL21.
- Expressed in E. coli for recombinant production.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Demonstrated chemotactic activity at 10-100 ng/mL (ED50 11.61 ng/mL).
- Lyophilized formulation: 20 mM PB, 150 mM NaCl, pH 7.4.
- Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C after reconstitution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.