Learn More
Medchemexpress LLC Exopolyphosphatase/P 100ug
SDP
Supplier:  Medchemexpress LLC HYP71516100UG
Encompass_Preferred
Recombinant exopolyphosphatase (PPX) from Escherichia coli supplied as a lyophilized powder with N-terminal His and SUMO tags. The enzyme hydrolyzes inorganic polyphosphate to release orthophosphate, preferring long-chain polyphosphates, and is provided at a laboratory scale quantity suitable for in vitro enzymatic assays.
  • Recombinant expression in E. coli with N-terminal His and SUMO tags.
  • Lyophilized powder for improved stability and storage.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Preferential activity toward long-chain polyphosphates.
  • Suitable for in vitro enzymatic assays and polyphosphate metabolism studies.
  • Available documentation includes datasheet, SDS, and COA for quality and handling.
Catalog No. 50-001-96394