Learn More
Medchemexpress LLC Exopolyphosphatase/P 50ug
SDP
Supplier:  Medchemexpress LLC HYP7150250UG
Encompass_Preferred
Recombinant exopolyphosphatase (EC 3.6.1.11) expressed in Escherichia coli with an N-terminal 6xHis tag, provided for research use in microgram quantities. The enzyme catalyzes processive hydrolysis of inorganic polyphosphate to orthophosphate and is intended for biochemical and enzymatic assays; SDS and a certificate of analysis are available.
  • Recombinant enzyme expressed in Escherichia coli with an N-terminal His tag.
  • Catalyzes processive hydrolysis of inorganic polyphosphate to orthophosphate.
  • Supplied in microgram-scale sizes, including a 50 μg option.
  • SDS and certificate of analysis available for quality and safety data.
  • Store under conditions specified on the certificate of analysis.
  • Suitable for biochemical assays and enzymology studies.
Catalog No. 50-001-96393