Learn More
System Biosciences LLC EXOQUICK PLASMA AND EXOSOME KITExoQuick Plasma prep and Exosome precipitation kit (5 ml ExoQuick plus 500 µL Thrombin > 500U/mL) = 75 Reactions
SDP
Supplier:  System Biosciences LLC EXOQ5TM1
Encompass_Preferred
ExoQuick Plasma Prep with Thrombin. When isolating exosomes from plasma, fibrinogen and fibrin can impede efficient recovery. By pre-treating plasma with thrombin, the fibrinogen can be converted to fibrin and easily pre-cleared by a simple centrifugation step. The resulting serum-like solution can now be treated with ExoQuick, original formulation, for quantitative exosome isolation that is compatible with high-throughput methods.The ExoQuick Plasma Prep with Thrombin Kit includes both ExoQuick and thrombin for efficient exosome isolation from plasma.
Catalog No. NC0567736