Learn More
System Biosciences LLC EXOQUICK-TC FOR TC MEDIA 50MLExoQuick-TC = 50 mL
SDP
Supplier:  System Biosciences LLC EXOTC50A1
Encompass_Preferred
Fast, scalable exosome isolation from tissue culture media. Need exosomes? SBI's ExoQuick-TC enables high-throughput, quantitative isolation of exosomes from low volumes (as little as 1 ml) of tissue culture media and certain biofluids (saliva, urine, follicular fluid, and breast milk). Note, to isolate exosomes from serum, plasma, or ascites fluid, use the original ExoQuick formulation (EXOQ5A-1 or EXOQ20A-1). Compatible with a wide variety of downstream applications, ExoQuick-TC is an effective and proven alternative to ultracentrifugation.
Catalog No. NC0169049