Learn More
Eurofins Discoverx PathHunter eXpress F2RL3 (PAR4) CHO-K1 β-Arrestin GPCR Assay, 2pl Kit. (93-0279E2CP0M)
Supplier:  Eurofins Discoverx 930279E2CP0M
Encompass
The PathHunter F2RL3 (PAR4) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. serine proteases) based activation of F2RL3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged F2RL3 and EA-tagged β-Arrestin-2. Activation of F2RL3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0160