Learn More
Eurofins Discoverx cAMP Hunter eXpress GPBAR1 CHO-K1 GPCR Assay, 10-Plate Kit. (95-0049E2CP2L)
Supplier:  Eurofins Discoverx 950049E2CP2L
Encompass
The cAMP Hunter GPBAR1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPBAR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPBAR1, a naturally Gs coupled GPCR. Upon activation of GPBAR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2458
Only null left
Add to Cart
Request a Quote