Learn More
Eurofins Discoverx PathHunter eXpress PDGFRa Functional Assay, 10-Plate Kit. (93-0823E3CP16L)
Supplier:  Eurofins Discoverx 930823E3CP16L
Encompass
The PathHunter eXpress PDGFRa Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated PDGFRa dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged PDGFRa and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1364
Add to Cart
Request a Quote