Learn More
Optimize Technologies EXP™2 In-Line Filter Holder (Fittings Included, for use w/EXP™2 Filter Cartridges)
SDP
Supplier:  Optimize Technologies 150405258
Encompass_Preferred
The EXP™2 Pre-Column and In-Line Hand-Tight Filters are ideal for protecting small-particle UHPLC columns and systems at extreme pressures. Such techniques analyze samples in the most demanding applications, decreasing the life of expensive hardware. EXP™2 Filters help extend the life of a UHPLC system and protect your investment without sacrificing performance. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet. EXP™2 Pre-Column Filter holders are available with Titanium Hybrid Ferrules for easy direct connection to any 10-32 port. The filter holder is a complete package and includes fittings to provide repeated zero-dead-volume connections.
Catalog No. 50-200-6490